Acrylonitrile, 99+%
Acrylonitrile, 99+%
Acrylonitrile, 99+%
Thermo Scientific Chemicals

Acrylonitrile, 99+%

CAS: 107-13-1 | C3H3N | 53.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 149630100
Price (USD)
210.65
Special offer
Online exclusive
Ends: 28-Dec-2026
248.00
Save 37.35
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
210.65
Special offer
Online exclusive
Ends: 28-Dec-2026
248.00
Save 37.35
Each
Chemical Identifiers
CAS107-13-1
IUPAC Nameprop-2-enenitrile
Molecular FormulaC3H3N
InChI KeyNLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILESC=CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Stabilizer35 to 45 ppm MEHQ
Water=<0.5 %
Color scale=<25 APHA
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only