trans-3-Hexene, 98%
trans-3-Hexene, 98%
trans-3-Hexene, 98%
Thermo Scientific Chemicals

trans-3-Hexene, 98%

CAS: 13269-52-8 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 149910051
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13269-52-8
IUPAC Name(3E)-hex-3-ene
Molecular FormulaC6H12
InChI KeyZQDPJFUHLCOCRG-AATRIKPKSA-N
SMILESCC\C=C\CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3935 to 1.3955 (20°C, 589 nm)
Assay>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only