Particle Certified HDPE Containers
Particle Certified HDPE Containers
Actual product may vary
Thermo Scientific™

Particle Certified HDPE Containers

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Certified
150-01WNLPM
also known as 150-01W/N/LPM
1000 mLLow Particulate, Low Metals
156-125WNLPM
also known as 156-125W/N/LPM
125 mLLow Particulate, Low Metals
156-125WNLP125 mLLow Particulate
157-250WNLPM
also known as 157-250W/N/LPM
250 mLLow Particulate, Low Metals
157-250WNLP250 mLLow Particulate
150-01WNLP1000 mLLow Particulate
Catalog number 150-01WNLPM
also known as 150-01W/N/LPM
Price (HKD)
-
Request A Quote
Capacity (Metric):
1000 mL
Certified:
Low Particulate, Low Metals

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:

  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity32 oz., 1000mL
CertifiedLow Particulate, Low Metals
Closure MaterialPolypropylene
Closure Size38-430 mm
Diameter (English)3.625 in.
Diameter (Metric)92.08 mm
Height (English)8.5 in.
Height (Metric)215.9 mm
No. per Case12
Product LineNalgene
Volume (English) Brim32 oz.
Volume (Metric) Brim1000mL
Workflow StepUpstream process solution storage. Upstream Validation. Upstream process solution storage
Capacity (English)32 oz.
Capacity (Metric)1000 mL
Certifications/ComplianceLow metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
Product TypeSquare Water Sampling Bottle
ShapeRound
Unit SizeCase of