4-Chlorobenzoyl chloride, 99+%
4-Chlorobenzoyl chloride, 99+%
4-Chlorobenzoyl chloride, 99+%
Thermo Scientific Chemicals

4-Chlorobenzoyl chloride, 99+%

CAS: 122-01-0 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 150190025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-01-0
IUPAC Name4-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyRKIDDEGICSMIJA-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to slightly colored
Appearance (Form)Liquid
Infrared spectrumConforms
Titration RCOCl>=98.50 %
Refractive index1.5770 to 1.5790 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only