4-chlorotoluene, 98%
4-chlorotoluene, 98%
4-chlorotoluene, 98%
Thermo Scientific Chemicals

4-chlorotoluene, 98%

CAS: 106-43-4 | C7H7Cl | 126.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number 150210010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-43-4
IUPAC Name1-chloro-4-methylbenzene
Molecular FormulaC7H7Cl
InChI KeyNPDACUSDTOMAMK-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5190 to 1.5230 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only