Diethyl phenylmalonate, 98%
Diethyl phenylmalonate, 98%
Diethyl phenylmalonate, 98%
Thermo Scientific Chemicals

Diethyl phenylmalonate, 98%

CAS: 83-13-6 | C13H16O4 | 236.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 150261000
Price (EUR)
26,70
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,70
Each
Chemical Identifiers
CAS83-13-6
IUPAC Name1,3-diethyl 2-phenylpropanedioate
Molecular FormulaC13H16O4
InChI KeyFGYDHYCFHBSNPE-UHFFFAOYSA-N
SMILESCCOC(=O)C(C(=O)OCC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Refractive index1.4900 to 1.4930 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only