Nunc™ IVF Dishes
Nunc™ IVF Dishes
Nunc™ IVF Dishes
Nunc™ IVF Dishes
Thermo Scientific™

Nunc™ IVF Dishes

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)Culture Area
15027054 mm14 mm21.5 cm2
15025535 mm11 mm8.8 cm2
15036088 mm15 mm56.7 cm2
Catalog number 150270
Price (USD)
651.65
Online exclusive
672.00
Save 20.35
Case of 400
Diameter (Metric):
54 mm
Height (Metric):
14 mm
Culture Area:
21.5 cm2
Price (USD)
651.65
Online exclusive
672.00
Save 20.35
Case of 400

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Product features for reliable results:

  • Facilitate gas exchange with vented lid
  • Available in 35mm, 60mm and 90mm sizes
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
DescriptionIVF Dish, 60mm, Lid w/Airvent
No. per Case400
No. per Pack10
Culture Area21.5 cm2
Diameter (Metric)54 mm
Height (Metric)14 mm
MaterialPolystyrene (USP Class VI)
SterilitySterile
Surface TreatmentUntreated
Unit SizeCase of 400