2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
Thermo Scientific Chemicals

2-Methyl-2-propen-1-ol, 98%

CAS: 513-42-8 | C4H8O | 72.11 g/mol
数量:
10 mL
100 mL
500 mL
製品番号(カタログ番号) 150295000
価格(JPY)
-
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS513-42-8
IUPAC Name2-methylprop-2-en-1-ol
Molecular FormulaC4H8O
InChI KeyBYDRTKVGBRTTIT-UHFFFAOYSA-N
SMILESCC(=C)CO
さらに表示
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only