2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
Thermo Scientific Chemicals

2-Methyl-2,4-pentanediol, 99%

CAS: 107-41-5 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 150340010
Price (USD)
47.50
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
47.50
Each
Chemical Identifiers
CAS107-41-5
IUPAC Name2-methylpentane-2,4-diol
Molecular FormulaC6H14O2
InChI KeySVTBMSDMJJWYQN-UHFFFAOYNA-N
SMILESCC(O)CC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only