Diphenylacetonitrile, 99+%
Diphenylacetonitrile, 99+%
Diphenylacetonitrile, 99+%
Thermo Scientific Chemicals

Diphenylacetonitrile, 99+%

CAS: 86-29-3 | C14H11N | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 150365000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS86-29-3
IUPAC Name2,2-diphenylacetonitrile
Molecular FormulaC14H11N
InChI KeyNEBPTMCRLHKPOB-UHFFFAOYSA-N
SMILESN#CC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point73°C to 76°C
GC>=99.0 %
Water=<0.2 % (K.F.)
Appearance (Color)White to cream to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only