4'-Hydroxyacetophenone, 98%
4'-Hydroxyacetophenone, 98%
4'-Hydroxyacetophenone, 98%
Thermo Scientific Chemicals

4'-Hydroxyacetophenone, 98%

CAS: 99-93-4 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 150405000
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS99-93-4
IUPAC Name1-(4-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O2
InChI KeyTXFPEBPIARQUIG-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point107°C to 111°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only