Ethyl cinnamate, 98% trans
Ethyl cinnamate, 98% trans
Ethyl cinnamate, 98% trans
Thermo Scientific Chemicals

Ethyl cinnamate, 98% trans

CAS: 103-36-6 | C11H12O2 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 150530025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-36-6
IUPAC Nameethyl (2E)-3-phenylprop-2-enoate
Molecular FormulaC11H12O2
InChI KeyKBEBGUQPQBELIU-CMDGGOBGSA-N
SMILESCCOC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(25/25°C) 1.045 to 1.048
Acid value=<1 mg KOH/g
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5580 to 1.5610 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only