L(-)-Malic acid, 99%
L(-)-Malic acid, 99%
L(-)-Malic acid, 99%
Thermo Scientific Chemicals

L(-)-Malic acid, 99%

CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 150591000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS97-67-6
IUPAC Name(2S)-2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-REOHCLBHSA-N
SMILESO[C@@H](CC(O)=O)C(O)=O
さらに表示
Sulfate (SO4)=<300 ppm
Chloride (Cl)=<100 ppm
Fumaric acid=<0.2 %
Appearance (Color)White
Appearance (Form)Crystalline powder or granules
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only