This product ships via ground from US northeast.
D(+)-Mannose, 99+%
D(+)-Mannose, 99+%
D(+)-Mannose, 99+%
Thermo Scientific Chemicals

D(+)-Mannose, 99+%

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 150600010
Price (USD)
908.65
Online exclusive
1,009.00
Save 100.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
908.65
Online exclusive
1,009.00
Save 100.35
Each
Chemical Identifiers
CAS3458-28-4
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<0.45 %
Heavy metals (as Pb)=<10 ppm
Specific optical rotation+13.5° to +14.9° (20°C, 589 nm) (c=10, water)
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only