1-Octanol, 99%, pure
1-Octanol, 99%, pure
1-Octanol, 99%, pure
Thermo Scientific Chemicals

1-Octanol, 99%, pure

CAS: 111-87-5 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 150630010
Price (USD)
87.65
Online Exclusive
97.30
Save 9.65 (10%)
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.30
Save 9.65 (10%)
Each
Chemical Identifiers
CAS111-87-5
IUPAC Nameoctan-1-ol
Molecular FormulaC8H18O
InChI KeyKBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILESCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only