Pyridoxine hydrochloride, 98+%, extra pure
Pyridoxine hydrochloride, 98+%, extra pure
Pyridoxine hydrochloride, 98+%, extra pure
Thermo Scientific Chemicals

Pyridoxine hydrochloride, 98+%, extra pure

CAS: 58-56-0 | C8H11NO3·HCl | 205.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 150772500
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
320.00
Save 47.35
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
320.00
Save 47.35
Each
Chemical Identifiers
CAS58-56-0
IUPAC Namehydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
Molecular FormulaC8H12ClNO3
InChI KeyZUFQODAHGAHPFQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
SolubilitySolution
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 % (On dry basis)
Loss on drying=<0.5 % (1 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only