Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer
Fisher Scientific™

Traceable™ Excursion-Trac™ Datalogging Thermometer

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
15081123
also known as FISHERTOPSELLING-15081123-EACH
1 Bottle Probe
15081126
also known as FISHERTOPSELLING-15081126-EACH
2 Probes, 5/16 in. dia. x 3/4 in. long
15081127
also known as FISHERTOPSELLING-15081127-EACH
2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
15081128
also known as FISHERTOPSELLING-15081128-EACH
1 Stainless-Steel Probe
15081129
also known as FISHERTOPSELLING-15081129-EACH
2 Stainless-Steel Probes
15079660
also known as FISHERTOPSELLING-15079660-EACH
1 Plastic Bottle Probe
15079661
also known as FISHERTOPSELLING-15079661-EACH
2 Plastic Bottle Probes
Catalog number 15081123
also known as FISHERTOPSELLING-15081123-EACH
Price (USD)
-
Includes:
1 Bottle Probe
Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01°C and maintains an accuracy of ±0.25°C across the entire temperature range.

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Datalogging Capacity: 525,600 temperature observations
  • Weight: 5 oz.

Hi/Lo Alarms and Time/Date Stamps

  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display

USB Data Transferring

  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location

Data-Logging Features

  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals

Bottle Probe Features (Cat. Nos. 15-081-125, -126, -128, 129 do not come with bottle probes)

  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Note: Units are designed to be used with either the provided AC adaptor or with batteries only. Batteries are designed to last for up to six months. If using battery-power only, the AC power adaptor must be plugged in to download any data.

Specifications
Accuracy (Temperature)±0.25°C
CalibrationCalibrated
Certifications/ComplianceISO 17025, A2LA, NIST
Data Memory525,600 Readings
Description1 Bottle Probe
For Use With (Application)Monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.
Includes1 Bottle Probe
InterfaceUSB
Length (English) Cable10 ft. (Cable)
Resolution0.01°
Response TimeUser-Defined Timing Interval: Yes
Sensor TypesTemperature-Buffered Bottle Sensor
Temperature Range (English)-58° to +158°F
Temperature Range (Metric)-50.00°C to +70.00°C
Temperature Resolution0.01°
Alarm TypesLED Indicator, Audible, Flashing LCD Segment
Display TypeLCD
Software TypeTraceableLIVE
TypeDatalogger
Unit SizeEach