2,5-Di-tert-butylhydroquinone, 99%
2,5-Di-tert-butylhydroquinone, 99%
2,5-Di-tert-butylhydroquinone, 99%
Thermo Scientific Chemicals

2,5-Di-tert-butylhydroquinone, 99%

CAS: 88-58-4 | C14H22O2 | 222.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 150840010
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Chemical Identifiers
CAS88-58-4
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Light beige to pink-beige
Melting point212°C to 218°C
Appearance (Form)Crystalline powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only