Isopropyl acetate, 99%, pure
Isopropyl acetate, 99%, pure
Isopropyl acetate, 99%, pure
Isopropyl acetate, 99%, pure
Thermo Scientific Chemicals

Isopropyl acetate, 99%, pure

CAS: 108-21-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Metal Drum
Catalog number 150860250
Price (USD)
757.65
Special offer
Online exclusive
Ends: 30-Jun-2026
891.00
Save 133.35 (15%)
Each
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
757.65
Special offer
Online exclusive
Ends: 30-Jun-2026
891.00
Save 133.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-21-4
IUPAC Namepropan-2-yl acetate
Molecular FormulaC5H10O2
InChI KeyJMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILESCC(C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3760 to 1.3780 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only