Crotonic acid, 98%
Crotonic acid, 98%
Crotonic acid, 98%
Thermo Scientific Chemicals

Crotonic acid, 98%

CAS: 107-93-7 | C4H6O2 | 86.09 g/mol
数量:
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 150870025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS107-93-7
IUPAC Name(2E)-but-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyLDHQCZJRKDOVOX-NSCUHMNNSA-N
SMILESC\C=C\C(O)=O
さらに表示
Appearance (Color)White to light yellow
Appearance (Form)Crystalline flakes or pellets
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
GC>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only