L(-)-3-Phenyllactic acid, 98%
L(-)-3-Phenyllactic acid, 98%
L(-)-3-Phenyllactic acid, 98%
Thermo Scientific Chemicals

L(-)-3-Phenyllactic acid, 98%

CAS: 20312-36-1 | C9H9O3 | 165.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 150920050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20312-36-1
IUPAC Name(2S)-2-hydroxy-3-phenylpropanoate
Molecular FormulaC9H9O3
InChI KeyVOXXWSYKYCBWHO-QMMMGPOBSA-M
SMILESO[C@@H](CC1=CC=CC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point121°C to 125°C
Specific optical rotation-17° to -20° (24°C, 589 nm) (c=1, methanol)
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
TLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only