3-Aminopropyltriethoxysilane, 99%
3-Aminopropyltriethoxysilane, 99%
3-Aminopropyltriethoxysilane, 99%
Thermo Scientific Chemicals

3-Aminopropyltriethoxysilane, 99%

CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 151085000
Price (EUR)
213,65
Online exclusive
237,00
Save 23,35 (10%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
213,65
Online exclusive
237,00
Save 23,35 (10%)
Each
Chemical Identifiers
CAS919-30-2
IUPAC Name(3-aminopropyl)triethoxysilane
Molecular FormulaC9H23NO3Si
InChI KeyWYTZZXDRDKSJID-UHFFFAOYSA-N
SMILESCCO[Si](CCCN)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
Color scale=<25 APHA
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only