Thermo Scientific™

Pierce™ Biotin-Binding Plate Sample Pack

Catalog number:  15115

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Thermo Scientific Pierce Biotin-Binding Plate Sample Pack is ideal for establishing which variety of streptavidin coated plate is best for particular assay involving biotinylated molecules.

The Biotin-Binding Plate Sample Pack helps in assay development. This sample pack contains streptavidin and NeutrAvidin Protein coated on microplates that are preblocked with either SuperBlock, Blocking Buffer or Blocker BSA. This variety allows researchers to determine which plate produces the best signal-to-noise ratio for a specific experimental system. Once optimal conditions are empirically determined, the plate that provides the best results can be purchased in bulk for the remainder of the study.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property: Affinity, High Capacity, Standard Capacity
Color: Clear
Detection Method: Colorimetric
Format: 96-well strip plate
Plate Blocking: Blocker™ BSA Buffer, SuperBlock™ Buffer
Plate Type: Detection Plate, ELISA, Immunoassay
Product Line: Pierce™
Product Size: 4 plates
Surface Coating: NeutrAvidin™ Protein, Streptavidin

Contents & storage

Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature.

Documents

Manuals & protocols