1,3-Dichlorobenzene, 98%
1,3-Dichlorobenzene, 98%
1,3-Dichlorobenzene, 98%
Thermo Scientific Chemicals

1,3-Dichlorobenzene, 98%

CAS: 541-73-1 | C6H4Cl2 | 147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
Catalog number 151180010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS541-73-1
IUPAC Name1,3-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyZPQOPVIELGIULI-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only