Pierce™ Streptavidin Coated Plates, Clear, 96-Well

Catalog number: 15124B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Streptavidin Coated Plates, Clear, 96-Well are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.

Features of Streptavidin Coated Plates, Clear, 96-Well:

• Approximate binding capacity is ~5 pmol biotin/well
• Easy and gentle immobilization of biotin-containing conjugates
• Low nonspecific binding
• No denaturing of the protein component of a conjugate upon binding
• Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
• Pre-blocked with your choice of Blocker BSA or SuperBlock Blocking Buffer

Streptavidin Coated Plates, Clear are available in 96-well and 384-well plate formats, pre-blocked with BSA or SuperBlock Blocking Buffers. Choose from clear, white and black plates for use with colorimetric (chromogenic), chemiluminescent and fluorescent detection methods, respectively.

Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.

Related Products
Pierce™ Streptavidin Coated Plates, Clear, 8-Well Strip
Pierce™ Streptavidin Coated Plates, Clear, 384-Well
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property: Affinity, Standard Capacity
Color: Clear
Detection Method: Colorimetric
Format: 96-well plate
Plate Blocking: SuperBlock™ Buffer
Plate Type: Detection Plate, ELISA, Immunoassay
Product Line: Pierce™
Product Size: Custom
Surface Coating: Streptavidin

Contents & storage

Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.

Documents

Manuals & protocols