P 211 Neutron Generator
Thermo Scientific™
P 211 Neutron Generator
Thermo Scientific™ P 211 is designed so that zero neutrons are produced between pulses.
Have Questions?
Número do catálogoDescription
1512521CP 211 Neutron Generator
Número do catálogo 1512521C
Preço (BRL)
-
Description:
P 211 Neutron Generator
Thermo Scientific™ P 211 is designed so that zero neutrons are produced between pulses. Assures that no neutrons are produced between pulses, which is critical for applications measuring neutron die away from the neutron burst.
This product cannot in the foreseeable future be offered for sale due to obsolete components which cannot be obtained. Thermo Fisher Scientific is still able to support existing systems with replacement neutron tubes and most repairs.
The features of P 211 are critical for applications that depend upon measuring the variations of neutron populations following the source burst. The P 211 neutron tube was originally designed by Sandia National Laboratories and incorporates the Sandia-designed “Zetatron” neutron tube.
  • Neutron die-away measurements
  • Pulses both accelerating voltage and ion source
  • No background between pulses
  • Analysis methods: Neutron die-away
  • Generator characteristics: Pulsed output (no background between pulses)
  • Primary elements detected: U, Pu
  • Pulse width: ∽10μsec
Recommended for:
  • Transuranic waste assay
  • Fissionable materials
Note:
Thermo is now the only manufacturer of the Zetatron neutron tube. Thermo supplies P 211 neutron generators as well as Zetatron neutron tubes to Sandia and other government and private users.
Especificações
DescriptionP 211 Neutron Generator
Item DescriptionP 211 Neutron Generator
Neutron Yield1 x 10&sup8; n/s
Outputs1 x 10&sup6; n/pulse (Neutron)
Safety FeaturesKeyed ON/OFF switch, user configurable interlocks
Typical Tube LifetimeUp to 500 hours or greater
Unit SizeEach