Bromocresol Green, sodium salt, pure, water soluble, indicator
Bromocresol Green, sodium salt, pure, water soluble, indicator
Bromocresol Green, sodium salt, pure, water soluble, indicator
Bromocresol Green, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals

Bromocresol Green, sodium salt, pure, water soluble, indicator

CAS: 67763-24-0 | C21H13Br4NaO5S | 720.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151320050
Price (EUR)
30,60
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,60
Each
Add to cart
Chemical Identifiers
CAS67763-24-0
IUPAC Namesodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular FormulaC21H13Br4NaO5S
InChI KeyHEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to dark blue to black
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrumComplies
Clarity of solution(0.1% in water) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only