Pierce™ Goat Anti-Rabbit IgG Antibody Coated Plates, Clear, 96-Well

Catalog number: 15135

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Goat Anti-Rabbit IgG Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates. These 96-well plates are clear for colorimetric assays.

Features of Goat Anti-Rabbit IgG Antibody Coated Plates:

• Prevents antibody denaturation as a result of direct binding to polystyrene
• Unlike Protein A or Protein G Plates, these plates bind only to target species
• Antibody-binding capacity is higher than direct adsorption onto polystyrene
• Pre-blocked to reduce nonspecific binding
• Binding Capacity: ~12 pmol IgG/well
• Activation level: 100 µL/well

Because these plates are IgG-specific, purified antibodies are not required. These plates may be used for direct, indirect, competitive or sandwich assays.

Related Products
Pierce™ Goat Anti-Rabbit IgG Antibody Coated Plates, White, 96-Well
Pierce™ Goat Anti-Rabbit IgG Antibody Coated Plates, Black, 96-Well
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property: Affinity, Standard Capacity
Color: Clear
Detection Method: Colorimetric
Format: 96-well plate
Plate Blocking: SuperBlock™ Buffer
Plate Type: Detection Plate, ELISA, Immunoassay
Product Line: Pierce™
Product Size: 5 plates
Surface Coating: Antibody

Contents & storage

Note: Plates are activated to 100 µL and supplied pre-blocked with SuperBlock® Blocking Buffer.

Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag and store desiccated at 4°C.

Documents

Manuals & protocols