Bromothymol Blue, pure, indicator
Bromothymol Blue, pure, indicator
Bromothymol Blue, pure, indicator
Thermo Scientific Chemicals

Bromothymol Blue, pure, indicator

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol
Have Questions?
Catalog NumberQuantityPackaging
151361000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151361000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS76-59-5
IUPAC Name3,3-bis[3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC27H28Br2O5S
InChI KeyNUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV spectrumComplies
Visual transition intervalfrom pH 6.0 (yellow) to pH 7.6 (blue)
Appearance (Color)Light pink to brown to purple
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only