Cresol Red, pure, Indicator grade
Cresol Red, pure, Indicator grade
Cresol Red, pure, Indicator grade
Acros Organics

Cresol Red, pure, Indicator grade

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1513800505 gGlass bottle
Catalog number 151380050
HSN 29349900
Price (INR)
5,999.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,999.00
Each
Chemical Identifiers
CAS1733-12-6
IUPAC Name3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H18O5S
InChI KeyOBRMNDMBJQTZHV-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific extinction E (1%/1cm)>=1500 (572 nm, in 0.01 N NaOH)
Appearance (Color)Dark green to red-brown
Infrared spectrumConforms
Solubility(0.1 % in 50 % ethanol) Clear
Visual transition intervalfrom pH 7.0 (orange) to pH 8.8 (purple)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only