Methyl Yellow, pure, indicator grade
Methyl Yellow, pure, indicator grade
Methyl Yellow, pure, indicator grade
Thermo Scientific Chemicals

Methyl Yellow, pure, indicator grade

CAS: 60-11-7 | C14H15N3 | 225.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 151405000
Price (EUR)
628,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
628,00
Each
Chemical Identifiers
CAS60-11-7
IUPAC NameN,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC14H15N3
InChI KeyJCYPECIVGRXBMO-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Loss on drying=<3 %
Appearance (Form)Powder
Visual transition intervalfrom pH 2.9 (red) to pH 4.0 (yellow)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only