Phenol Red, pure, indicator
Phenol Red, pure, indicator
Phenol Red, pure, indicator
Thermo Scientific Chemicals

Phenol Red, pure, indicator

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151430050
Price (EUR)
34,65
Online exclusive
37,80
Save 3,15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,65
Online exclusive
37,80
Save 3,15
Each
Chemical Identifiers
CAS143-74-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H14O5S
InChI KeyBELBBZDIHDAJOR-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max555 to 561 nm (in buffer)
Solubility(0.1 % in 95% ethanol) Clear soluble
Appearance (Color)Red
Infrared spectrumConforms
Visual transition intervalfrom pH 6.8 (yellow) to pH 8.2 (red)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only