Phenol Red, pure, indicator
Phenol Red, pure, indicator
Phenol Red, pure, indicator
Acros Organics

Phenol Red, pure, indicator

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 151430250
HSN 29349900
Price (INR)
8,230.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,230.00
Each
Chemical Identifiers
CAS143-74-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H14O5S
InChI KeyBELBBZDIHDAJOR-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV Lambda max555 to 561 nm (in buffer)
Solubility(0.1 % in 95% ethanol) Clear soluble
Appearance (Color)Red
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only