Thymol Blue, pure, indicator
Thymol Blue, pure, indicator
Thymol Blue, pure, indicator
Thermo Scientific Chemicals

Thymol Blue, pure, indicator

CAS: 76-61-9 | C27H30O5S | 466.592 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151450250
Price (EUR)
105,65
Online exclusive
117,00
Save 11,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
105,65
Online exclusive
117,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS76-61-9
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC27H30O5S
InChI KeyPRZSXZWFJHEZBJ-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Solubility(0.1 % in ethanol) Clear
Specific extinction E (1%/1cm)>=700 (596 nm, in 0.01 N NaOH)
Appearance (Color)Dark green to dark brown
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only