4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
Thermo Scientific Chemicals

4-(4-Nitrophenylazo)resorcinol, 90%

CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 151511000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS74-39-5
IUPAC Name3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC12H9N3O4
InChI KeyBIFFKKLAAIGZBV-UHFFFAOYSA-N
SMILESOC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Orange-red to red
HPLC>=88 %
Appearance (Form)Powder
Specific extinction E (1%/1cm)>=1400 (554 to 559 nm, 1N NaOH)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only