Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
Have Questions?
Change view
Catalog Number
Color
Format
Detection Method
15152
White
Standard
Chemiluminescence
15150
Clear
Strip Plate
Colorimetric
15153
Black
Standard
Fluorescence
3 Options
Catalog number 15152
Price (USD)
231.65
Online exclusive
252.00
Save 20.35
Each
Color:
White
Format:
Standard
Detection Method:
Chemiluminescence
Request bulk or custom format
Price (USD)
231.65
Online exclusive
252.00
Save 20.35
Each
Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights • Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls • Pre-blocked to reduce nonspecific binding
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Covalent
Detection MethodChemiluminescence
FormatStandard
No. per Pack5 Plates
Plate BlockingBlocker™ BSA Buffer
Product TypeDetection Plate, Immunoassay, ELISA
Quantity5 plates
Volume (Metric) Well100 μL
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentMaleimide
Unit SizeEach
Contents & Storage
Note: Plates are supplied pre-blocked with bovine serum albumin.
Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.