Thiophosgene, 85%
Thiophosgene, 85%
Thiophosgene, 85%
Thermo Scientific Chemicals

Thiophosgene, 85%

CAS: 463-71-8 | CCl2S | 114.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 151640250
Price (EUR)
44,65
Online exclusive
49,30
Save 4,65
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
44,65
Online exclusive
49,30
Save 4,65
Each
Chemical Identifiers
CAS463-71-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange-red to red
GC>=80 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only