1-Adamantanamine hydrochloride, 99+%
1-Adamantanamine hydrochloride, 99+%
1-Adamantanamine hydrochloride, 99+%
Thermo Scientific Chemicals

1-Adamantanamine hydrochloride, 99+%

CAS: 665-66-7 | C10H18ClN | 187.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 151671000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS665-66-7
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only