4,4'-Difluorobenzophenone, 99%
4,4'-Difluorobenzophenone, 99%
4,4'-Difluorobenzophenone, 99%
Thermo Scientific Chemicals

4,4'-Difluorobenzophenone, 99%

CAS: 345-92-6 | C13H8F2O | 218.20 g/mol
Catalog NumberQuantityPackaging
151791000100 gGlass bottle
Catalog number 151791000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS345-92-6
IUPAC Namebis(4-fluorophenyl)methanone
Molecular FormulaC13H8F2O
InChI KeyLSQARZALBDFYQZ-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point106°C to 109°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only