1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
Acros Organics

1,1'-Carbonyldiimidazole, 97%

CAS: 530-62-1 | C7H6N4O | 162.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
151811000100 gGlass bottle
Catalog number 151811000
HSN 29332990
Price (INR)
48,536.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
48,536.00
Each
Chemical Identifiers
CAS530-62-1
IUPAC Name1-(1H-imidazole-1-carbonyl)-1H-imidazole
Molecular FormulaC7H6N4O
InChI KeyPFKFTWBEEFSNDU-UHFFFAOYSA-N
SMILESO=C(N1C=CN=C1)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point111°C to 120°C
Titration with HClO496.0 to 104.0 %
Imidazole=<3 % (NMR)
Appearance (Color)White to light yellow to beige
Appearance (Form)Powder or crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only