1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
Thermo Scientific Chemicals

1,1'-Carbonyldiimidazole, 97%

CAS: 530-62-1 | C7H6N4O | 162.15 g/mol
数量:
10 g
25 g
100 g
500 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 151815000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS530-62-1
IUPAC Name1-(1H-imidazole-1-carbonyl)-1H-imidazole
Molecular FormulaC7H6N4O
InChI KeyPFKFTWBEEFSNDU-UHFFFAOYSA-N
SMILESO=C(N1C=CN=C1)N1C=CN=C1
さらに表示
Melting point111°C to 120°C
Titration with HClO496.0 to 104.0 %
Imidazole=<3 % (NMR)
Appearance (Color)White to light yellow to beige
Appearance (Form)Powder or crystalline powder or crystals
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only