Search
General description
• 2-Thiouracil (Thiouracil or 4-Hydroxy-2-mercaptopyrimidine) is a nucleotide analog with a C-2 thioxo substitution
• 2-Thiouracil is a melanocyte suppressor and thyroid inhibitor
Applications
• 2-Thiouracil can be used to inhibit nitric oxide synthase and suppresses nitric oxide signaling in vitro
• 2-Thiouracil can be used for labeling of RNA in vitro
• 2-Thiouracil is intended for research applications only
Literature references
Yang, W.; Shen, Z.; Wen, S.; Wang, W.; Hu, M. Mechanisms of multiple neurotransmitters in the effects of Lycopene on brain injury induced by Hyperlipidemia. Lipids Health Dis. 2018, 17(1), 13. Erratum in: Lipids Health Dis. 2018, 17 (1), 65.
Wauters, J.; Van, Meulebroek L.; Fichant, E.; Delahaut, P.; Vanhaecke, L. Discrimination between Synthetically Administered and Endogenous Thiouracil Based on Monitoring of Urine, Muscle, and Thyroid Tissue: An in Vivo Study in Young and Adult Bovines. J Agric Food Chem. 2017, 65(30), 6231-6239.