4-Clorobenzonitrilo, 99 %, Thermo Scientific Chemicals
4-Clorobenzonitrilo, 99 %, Thermo Scientific Chemicals
4-Clorobenzonitrilo, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Clorobenzonitrilo, 99 %, Thermo Scientific Chemicals

CAS: 623-03-0 | C7H4ClN | 137.57 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de plástico
Número de catálogo 151871000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS623-03-0
IUPAC Name4-chlorobenzonitrile
Molecular FormulaC7H4ClN
InChI KeyGJNGXPDXRVXSEH-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C#N
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Flakes and/or chunks
Melting point91°C to 94°C
Water=<0.1 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only