2-Bromo-4'-methoxyacetophenone, 98%
2-Bromo-4'-methoxyacetophenone, 98%
2-Bromo-4'-methoxyacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-4'-methoxyacetophenone, 98%

CAS: 2632-13-5 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 151901000
Price (USD)
352.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
352.00
Each
Chemical Identifiers
CAS2632-13-5
IUPAC Name2-bromo-1-(4-methoxyphenyl)ethan-1-one
Molecular FormulaC9H9BrO2
InChI KeyXQJAHBHCLXUGEP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
GC>=97.5 %
Melting point69°C to 72°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only