2,2,6,6-Tetramethylpiperidine, 98%
2,2,6,6-Tetramethylpiperidine, 98%
2,2,6,6-Tetramethylpiperidine, 98%
Thermo Scientific Chemicals

2,2,6,6-Tetramethylpiperidine, 98%

CAS: 768-66-1 | C9H19N | 141.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 151950250
Price (EUR)
114,65
Online exclusive
127,00
Save 12,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
114,65
Online exclusive
127,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS768-66-1
IUPAC Name2,2,6,6-tetramethylpiperidine
Molecular FormulaC9H19N
InChI KeyRKMGAJGJIURJSJ-UHFFFAOYSA-N
SMILESCC1(C)CCCC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow to light brown
Refractive index1.4425 to 1.4465 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only