96 Well Plate, Poly-D-Lysine Coated Surface, Pack of 5
Thermo Scientific™
96 Well Plate, Poly-D-Lysine Coated Surface, Pack of 5
Color:Clear; Lid:With Lid
Have Questions?
Catalog NumberQuantity
1520395/Pk., 20/Cs.
Catalog number 152039
Price (USD)
393.00
Case of 20
Quantity:
5/Pk., 20/Cs.
Price (USD)
393.00
Case of 20
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
  • Aseptically Manufactured
Specifications
Array8 x 12
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorClear
Description96F well plate, Poly-D-Lysine, clear, with lid
Packaging5/Pack, 20/Case
Product LineNunc
Quantity5/Pk., 20/Cs.
Surface TreatmentPoly-D-Lysine
Well ShapeFlat
MaterialPolystyrene
No. of Wells96
SterilityAseptically Manufactured
TypeMicroplate
Volume (Metric) Well400 μL
Unit SizeCase of 20
Frequently asked questions (FAQs)
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.