Trimethyl 1,3,5-benzenetricarboxylate, 99%
Trimethyl 1,3,5-benzenetricarboxylate, 99%
Trimethyl 1,3,5-benzenetricarboxylate, 99%
Thermo Scientific Chemicals

Trimethyl 1,3,5-benzenetricarboxylate, 99%

CAS: 2672-58-4 | C12H12O6 | 252.22 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 152071000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2672-58-4
IUPAC Name1,3,5-trimethyl benzene-1,3,5-tricarboxylate
Molecular FormulaC12H12O6
InChI KeyRGCHNYAILFZUPL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
さらに表示
Appearance (Form)Crystalline powder or crystals
Melting point143°C to 147°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only