Trichloroacetic acid, 99%, extra pure
Trichloroacetic acid, 99%, extra pure
Trichloroacetic acid, 99%, extra pure
Thermo Scientific Chemicals

Trichloroacetic acid, 99%, extra pure

CAS: 76-03-9 | C2HCl3O2 | 163.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 152130010
Price (USD)
168.65
Special offer
Online exclusive
Ends: 28-Dec-2026
198.00
Save 29.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 28-Dec-2026
198.00
Save 29.35
Each
Chemical Identifiers
CAS76-03-9
IUPAC Nametrichloroacetic acid
Molecular FormulaC2HCl3O2
InChI KeyYNJBWRMUSHSURL-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Sulfuric acid=<0.3 %
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only