2-Bromo-3-pyridinol, 99%
2-Bromo-3-pyridinol, 99%
2-Bromo-3-pyridinol, 99%
Thermo Scientific Chemicals

2-Bromo-3-pyridinol, 99%

CAS: 6602-32-0 | C5H4BrNO | 174.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 152170050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6602-32-0
IUPAC Name2-bromopyridin-3-ol
Molecular FormulaC5H4BrNO
InChI KeyYKHQFTANTNMYPP-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
Appearance (Color)Grey to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only