Distilled Water
Gibco™

Distilled Water

막여과, endotoxin 검사 완료.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
100 mL
500 mL
1000 mL
10 x 500 mL
20 x 100 mL
6 x 1 L
카탈로그 번호 15230170
제품 가격(KRW)
19,000
온라인 행사
Ends: 30-Sep-2026
21,000
할인액 2,000 (10%)
Each
수량:
100 mL
Customize this product
제품 가격(KRW)
19,000
온라인 행사
Ends: 30-Sep-2026
21,000
할인액 2,000 (10%)
Each
막여과, endotoxin 검사 완료.
For Research Use Only. Not for use in diagnostic procedures.
사양
CAS7732-18-5
화학물질 이름 또는 재질Dihydrogen monoxide
포장Bottle
정제 방법Membrane-Filtered
권장 스토리지Storage conditions: 15 – 30°C, Shipping conditions: Ambient, Shelf life: 24 months from date of manufacture
유통 기한24 Months
배송 조건Room Temperature
멸균Sterile-filtered
치료Not DEPC-Treated
등급Cell Culture
분자량(g/mol)18.02 g/mol
수량100 mL
pH6 to 8
Unit SizeEach

자주 묻는 질문(FAQ)

Is Distilled Water (Cat. No. 15230147) RNAse-free?

Distilled Water (Cat. No. 15230147) is not RNAse-free. Instead, we would recommend using UltraPure DNase/RNase-Free Distilled Water (Cat. No. 10977049) to have it DNase/RNase-free.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I get the conductivity measurement for Distilled Water?

We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.

Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.